Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.4450 in Dia, Steel
Catalog No. 5000530885
Encompass
This Class ZZ Minus Plug & Pin Gage features a precise 0.4450 inch diameter. Constructed from durable steel, it is designed for accurate measurement and inspection tasks.
  • Features a 0.4450 inch diameter measurement.
  • Provides a tolerance of 0.000200 "
  • Made from durable steel for longevity.
  • Manufactured in China.
Catalog No. 50-005-30885 Supplier MSC Supplier No. 78376415
Add to Cart
Add to Cart
This Class ZZ Minus Plug & Pin Gage features a precise 0.4450 inch diameter. Constructed from durable steel, it is designed for accurate measurement and inspection tasks.
  • Features a 0.4450 inch diameter measurement.
  • Provides a tolerance of 0.000200 "
  • Made from durable steel for longevity.
  • Manufactured in China.