Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.4480 in, Black Oxide
Catalog No. 5000527989
Encompass
The SPI Class ZZ Minus Plug & Pin Gage is a precision measurement tool designed for accurate verification of hole sizes. Featuring a 0.4480 inch diameter and a durable black oxide finish, this gage provides reliable performance with a tolerance of 0.0002 inches.
  • Precision measurement tool for hole verification.
  • 0.4480 inch diameter.
  • Class ZZ minus tolerance.
  • Durable black oxide finish.
  • Tolerance of 0.0002 inches.
Catalog No. 50-005-27989 Supplier MSC Supplier No. 56247976
The SPI Class ZZ Minus Plug & Pin Gage is a precision measurement tool designed for accurate verification of hole sizes. Featuring a 0.4480 inch diameter and a durable black oxide finish, this gage provides reliable performance with a tolerance of 0.0002 inches.
  • Precision measurement tool for hole verification.
  • 0.4480 inch diameter.
  • Class ZZ minus tolerance.
  • Durable black oxide finish.
  • Tolerance of 0.0002 inches.