Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.4570 in Diameter, Steel
Catalog No. 5000530761
Encompass
This SPI Class ZZ Minus Plug & Pin Gage is a precision instrument designed for accurate measurement of internal diameters. Manufactured from steel, it features a diameter of 0.4570 inches and a tight tolerance of 0.000200 inches, making it suitable for demanding quality control and inspection applications.
  • Precision ground to 0.4570 in diameter
  • Class ZZ Minus tolerance for critical measurements
  • Durable steel construction
  • Ideal for verifying hole sizes and depths
Catalog No. 50-005-30761 Supplier MSC Supplier No. 78376530
This SPI Class ZZ Minus Plug & Pin Gage is a precision instrument designed for accurate measurement of internal diameters. Manufactured from steel, it features a diameter of 0.4570 inches and a tight tolerance of 0.000200 inches, making it suitable for demanding quality control and inspection applications.
  • Precision ground to 0.4570 in diameter
  • Class ZZ Minus tolerance for critical measurements
  • Durable steel construction
  • Ideal for verifying hole sizes and depths