Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.4600 in Diameter, 0.0002 in Tolerance
Catalog No. 5000528978
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Manufactured from durable steel, it features a 0.4600 inch diameter with a tight 0.0002 inch tolerance, ensuring accuracy for critical inspections.
  • Precise 0.4600 inch diameter measurement
  • Tight 0.0002 inch tolerance for accurate gauging
  • Constructed from durable steel for longevity
  • Class ZZ minus designation for specific gauging requirements
Catalog No. 50-005-28978 Supplier MSC Supplier No. 78376563
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Manufactured from durable steel, it features a 0.4600 inch diameter with a tight 0.0002 inch tolerance, ensuring accuracy for critical inspections.
  • Precise 0.4600 inch diameter measurement
  • Tight 0.0002 inch tolerance for accurate gauging
  • Constructed from durable steel for longevity
  • Class ZZ minus designation for specific gauging requirements