Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.4740 in Diameter
Catalog No. 5000530488
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a 0.4740 inch diameter and is constructed from durable steel.
  • Diameter of measurement of 0.4740 in.
  • Precision tolerance of 0.000200 in (0.0002 mm).
  • Constructed from durable steel.
  • Manufactured in China.
Catalog No. 50-005-30488 Supplier MSC Supplier No. 78376704
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a 0.4740 inch diameter and is constructed from durable steel.
  • Diameter of measurement of 0.4740 in.
  • Precision tolerance of 0.000200 in (0.0002 mm).
  • Constructed from durable steel.
  • Manufactured in China.