Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.4800 in Diameter, Steel
Catalog No. 5000528737
Encompass
This SPI Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection of bore diameters. Crafted from durable steel, it ensures accuracy for quality control and manufacturing processes.
  • Provides a diameter of 0.4800 inches
  • Features a tight tolerance of 0.000200 inches
  • Constructed from steel for durability and reliability
  • Manufactured as a Class ZZ Minus gage
Catalog No. 50-005-28737 Supplier MSC Supplier No. 78376761
This SPI Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection of bore diameters. Crafted from durable steel, it ensures accuracy for quality control and manufacturing processes.
  • Provides a diameter of 0.4800 inches
  • Features a tight tolerance of 0.000200 inches
  • Constructed from steel for durability and reliability
  • Manufactured as a Class ZZ Minus gage