Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.4980 in, Steel
Catalog No. 5000511615
Encompass
This plug and pin gage is designed for precise measurement and quality control applications. Made from durable steel, it features a 0.4980 in diameter with a Class ZZ Minus tolerance of 0.000200 in, ensuring high accuracy for inspecting bore diameters.
  • Designed for precise measurement
  • 0.4980 in diameter
  • 0.000200 in tolerance
  • Made from steel
  • Class ZZ minus for accurate inspections
Catalog No. 50-005-11615 Supplier MSC Supplier No. 78376944
This plug and pin gage is designed for precise measurement and quality control applications. Made from durable steel, it features a 0.4980 in diameter with a Class ZZ Minus tolerance of 0.000200 in, ensuring high accuracy for inspecting bore diameters.
  • Designed for precise measurement
  • 0.4980 in diameter
  • 0.000200 in tolerance
  • Made from steel
  • Class ZZ minus for accurate inspections