Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.5070 in Dia, 0.0002 in Tolerance
Catalog No. 5000531513
Encompass
This plug and pin gage is designed for precise measurement applications. Made from durable steel, it features a 0.5070 inch diameter and a tolerance of 0.0002 inch, adhering to Class ZZ Minus standards.
  • Designed for precise measurement applications.
  • Constructed from steel for durability.
  • Features a 0.5070 inch diameter.
  • Provides a 0.0002 inch tolerance.
  • Adheres to Class ZZ Minus gauging standards.
Catalog No. 50-005-31513 Supplier MSC Supplier No. 78377033
This plug and pin gage is designed for precise measurement applications. Made from durable steel, it features a 0.5070 inch diameter and a tolerance of 0.0002 inch, adhering to Class ZZ Minus standards.
  • Designed for precise measurement applications.
  • Constructed from steel for durability.
  • Features a 0.5070 inch diameter.
  • Provides a 0.0002 inch tolerance.
  • Adheres to Class ZZ Minus gauging standards.