Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.5110 in Diameter, Black Oxide
Catalog No. 5000531777
Encompass
This SPI Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a 0.5110 inch diameter and a durable black oxide coating, making it suitable for a variety of industrial and scientific uses.
  • Precise measurement with a 0.5110 inch diameter.
  • Features a tight tolerance of 0.000200 "
  • Equipped with a black oxide coating for enhanced durability and corrosion resistance.
Catalog No. 50-005-31777 Supplier MSC Supplier No. 56248602
This SPI Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a 0.5110 inch diameter and a durable black oxide coating, making it suitable for a variety of industrial and scientific uses.
  • Precise measurement with a 0.5110 inch diameter.
  • Features a tight tolerance of 0.000200 "
  • Equipped with a black oxide coating for enhanced durability and corrosion resistance.