Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.5270 in Diameter, Steel
Supplier:  MSC 78377231
Encompass
This is an SPI Class ZZ Minus Plug & Pin Gage designed for precise measurement. It features a specific diameter and tolerance, making it suitable for applications requiring high accuracy.
  • Measures with a diameter of 0.5270 in
  • Maintains a tolerance of 0.000200 in
  • Constructed from durable steel
  • Manufactured in China
Catalog No. 50-005-28708