Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.5600 inch, Black Oxide
Catalog No. 5000531744
Encompass
This plug and pin gage is designed for precise measurement applications, featuring a 0.5600 inch diameter and a durable black oxide finish. It is classified as Class ZZ Minus, ensuring high accuracy with a tolerance of 0.000200 "
  • Class ZZ Minus for high accuracy
  • 0.5600 inch diameter for specific measurements
  • Black oxide finish for durability
  • Tolerance of 0.000200 inch for precision
Catalog No. 50-005-31744 Supplier MSC Supplier No. 56249097
This plug and pin gage is designed for precise measurement applications, featuring a 0.5600 inch diameter and a durable black oxide finish. It is classified as Class ZZ Minus, ensuring high accuracy with a tolerance of 0.000200 "
  • Class ZZ Minus for high accuracy
  • 0.5600 inch diameter for specific measurements
  • Black oxide finish for durability
  • Tolerance of 0.000200 inch for precision