Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.5830 in Dia, Steel
Catalog No. 5000530838
Encompass
This Class ZZ Minus Plug & Pin Gage features a precise 0.5830 inch diameter. It is constructed from durable steel and offers a tight tolerance of 0.000200 inches, making it suitable for accurate measurement applications.
  • Features a precise 0.5830 inch diameter for accurate measurements.
  • Maintains a tight tolerance of 0.000200 inches (0.0002 mm).
  • Constructed from durable steel for long-lasting performance.
Catalog No. 50-005-30838 Supplier MSC Supplier No. 78377793
This Class ZZ Minus Plug & Pin Gage features a precise 0.5830 inch diameter. It is constructed from durable steel and offers a tight tolerance of 0.000200 inches, making it suitable for accurate measurement applications.
  • Features a precise 0.5830 inch diameter for accurate measurements.
  • Maintains a tight tolerance of 0.000200 inches (0.0002 mm).
  • Constructed from durable steel for long-lasting performance.