Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.5860 in Diameter, Black Oxide
Supplier:  MSC 56249352
Encompass
This Class ZZ Minus plug and pin gage is designed for precise measurement applications. It features a 0.5860 in diameter with a black oxide finish, ensuring durability and accurate gaging for various industrial and laboratory tasks.
  • Designed for precise measurement applications.
  • Features a 0.5860 in diameter.
  • Class ZZ Minus for specific gaging requirements.
  • Black oxide finish for enhanced durability.
  • Provides a tolerance of 0.000200 in.
  • Manufactured in China.
Catalog No. 50-005-30111