Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.591 inch Diameter, Black Oxide
Catalog No. 5000531230
Encompass
This pin gage is designed for precise measurement applications. It features a black oxide coating for enhanced corrosion prevention and visual indication of wear, ensuring prolonged accuracy. Constructed from hardened drill rod, this gage offers durability and reliable performance.
  • Pin size is 0.591 inch
  • Features a black oxide coating to prevent corrosion and indicate wear
  • Constructed from hardened drill rod with RC 60-62 hardness
  • Overall length is 2 inch
Catalog No. 50-005-31230 Supplier MSC Supplier No. 56249402
This pin gage is designed for precise measurement applications. It features a black oxide coating for enhanced corrosion prevention and visual indication of wear, ensuring prolonged accuracy. Constructed from hardened drill rod, this gage offers durability and reliable performance.
  • Pin size is 0.591 inch
  • Features a black oxide coating to prevent corrosion and indicate wear
  • Constructed from hardened drill rod with RC 60-62 hardness
  • Overall length is 2 inch