Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.6080" Dia, Steel
Catalog No. 5000528730
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Constructed from durable steel, it features a diameter of 0.6080" and a tight tolerance of 0.000200" for accurate dimensional inspection.
  • Class ZZ minus plug and pin gage
  • Diameter of 0.6080"
  • Tolerance of 0.000200"
  • Made from steel
  • Suitable for precise measurement applications
Catalog No. 50-005-28730 Supplier MSC Supplier No. 78378049
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Constructed from durable steel, it features a diameter of 0.6080" and a tight tolerance of 0.000200" for accurate dimensional inspection.
  • Class ZZ minus plug and pin gage
  • Diameter of 0.6080"
  • Tolerance of 0.000200"
  • Made from steel
  • Suitable for precise measurement applications