Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.6130 in Diameter, Steel
Catalog No. 5000531065
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Made from durable steel, it features a 0.6130 inch diameter with a tight tolerance of 0.0002 inches, ensuring accuracy and reliability for quality control and inspection tasks.
  • Precise measurement for quality control
  • Durable steel construction
  • 0.6130 inch diameter for specific applications
  • Tight tolerance of 0.0002 inches ensures accuracy
Catalog No. 50-005-31065 Supplier MSC Supplier No. 78378098
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Made from durable steel, it features a 0.6130 inch diameter with a tight tolerance of 0.0002 inches, ensuring accuracy and reliability for quality control and inspection tasks.
  • Precise measurement for quality control
  • Durable steel construction
  • 0.6130 inch diameter for specific applications
  • Tight tolerance of 0.0002 inches ensures accuracy