Learn More
MSC SPI Plug and Pin Gage, Class ZZ Minus, 0.6480 in Diameter
Catalog No. 5000530060
Encompass
This Class ZZ Minus Plug and Pin Gage is designed for precise measurement applications. Constructed from durable steel, it features a 0.6480 in diameter and a strict tolerance of 0.000200 in, ensuring accuracy and reliability in various industrial and laboratory settings.
  • Class ZZ minus
  • 0.6480 in diameter
  • 0.000200 in tolerance
  • Made from steel
Catalog No. 50-005-30060 Supplier MSC Supplier No. 78378445
This Class ZZ Minus Plug and Pin Gage is designed for precise measurement applications. Constructed from durable steel, it features a 0.6480 in diameter and a strict tolerance of 0.000200 in, ensuring accuracy and reliability in various industrial and laboratory settings.
  • Class ZZ minus
  • 0.6480 in diameter
  • 0.000200 in tolerance
  • Made from steel