Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.682 inch Diameter, Black Oxide
Catalog No. 5000528738
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precision measurements, featuring a 0.682 inch diameter and a durable black oxide finish. It offers a tolerance of 0.000200 inch for accurate gauging.
  • Black oxide coating
  • 0.000200 inch tolerance
  • 0.0002 mm tolerance
Catalog No. 50-005-28738 Supplier MSC Supplier No. 78378783
This Class ZZ Minus Plug & Pin Gage is designed for precision measurements, featuring a 0.682 inch diameter and a durable black oxide finish. It offers a tolerance of 0.000200 inch for accurate gauging.
  • Black oxide coating
  • 0.000200 inch tolerance
  • 0.0002 mm tolerance