Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.6880 in Diameter, Steel
Catalog No. 5000529011
Encompass
This gage is designed for precise measurement applications. It is a Class ZZ Minus plug and pin gage, manufactured from durable steel to ensure accuracy and longevity in demanding environments.
  • Constructed from steel
  • Features a diameter of 0.6880 in
  • Provides a precise tolerance of 0.000200 in
  • Class ZZ minus for specific measurement requirements
Catalog No. 50-005-29011 Supplier MSC Supplier No. 78378841
Add to Cart
Add to Cart
This gage is designed for precise measurement applications. It is a Class ZZ Minus plug and pin gage, manufactured from durable steel to ensure accuracy and longevity in demanding environments.
  • Constructed from steel
  • Features a diameter of 0.6880 in
  • Provides a precise tolerance of 0.000200 in
  • Class ZZ minus for specific measurement requirements