Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.7000 in Dia, Black Oxide
Catalog No. 5000530113
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement. It features a 0.7000 in diameter and a black oxide finish, ensuring durability and accurate performance in various applications.
  • Features a precise 0.7000 in diameter.
  • Provides Class ZZ Minus tolerance for high accuracy.
  • Coated with a durable black oxide finish.
  • Offers a tight tolerance of 0.000200 in.
Catalog No. 50-005-30113 Supplier MSC Supplier No. 56250491
Add to Cart
Add to Cart
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement. It features a 0.7000 in diameter and a black oxide finish, ensuring durability and accurate performance in various applications.
  • Features a precise 0.7000 in diameter.
  • Provides Class ZZ Minus tolerance for high accuracy.
  • Coated with a durable black oxide finish.
  • Offers a tight tolerance of 0.000200 in.