Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.7000 in Diameter, Steel
Catalog No. 5000528982
Encompass
This plug and pin gage is designed for precise measurement applications. Featuring a 0.7000 inch diameter and a Class ZZ Minus tolerance, it provides accurate dimensional verification. Constructed from durable steel, this gage is suitable for various industrial and laboratory settings.
  • Designed for precise measurement applications.
  • Features a 0.7000 inch diameter.
  • Offers a Class ZZ Minus tolerance of 0.0002 "
  • Made from durable steel for longevity.
  • Provides accurate dimensional verification.
Catalog No. 50-005-28982 Supplier MSC Supplier No. 78378965
Add to Cart
Add to Cart
This plug and pin gage is designed for precise measurement applications. Featuring a 0.7000 inch diameter and a Class ZZ Minus tolerance, it provides accurate dimensional verification. Constructed from durable steel, this gage is suitable for various industrial and laboratory settings.
  • Designed for precise measurement applications.
  • Features a 0.7000 inch diameter.
  • Offers a Class ZZ Minus tolerance of 0.0002 "
  • Made from durable steel for longevity.
  • Provides accurate dimensional verification.