Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.7180 in, Steel
Catalog No. 5000528717
Encompass
This plug and pin gage is designed for precise measurement applications. It is a Class ZZ Minus type, ensuring high accuracy, and is constructed from durable steel for longevity.
  • Diameter of measurement: 0.7180 in
  • Tolerance: 0.000200 in
  • Made from steel
Catalog No. 50-005-28717 Supplier MSC Supplier No. 78379146
This plug and pin gage is designed for precise measurement applications. It is a Class ZZ Minus type, ensuring high accuracy, and is constructed from durable steel for longevity.
  • Diameter of measurement: 0.7180 in
  • Tolerance: 0.000200 in
  • Made from steel