Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.7310 in, Steel
Catalog No. 5000516343
Encompass
This plug and pin gage is designed for precise measurement applications. Manufactured from durable steel, it features a Class ZZ Minus tolerance and a measurement diameter of 0.7310 inches.
  • Precise 0.7310 in measurement diameter
  • Constructed from steel
  • Class ZZ Minus tolerance
  • Tight tolerance of 0.000200 in
Catalog No. 50-005-16343 Supplier MSC Supplier No. 78379278
This plug and pin gage is designed for precise measurement applications. Manufactured from durable steel, it features a Class ZZ Minus tolerance and a measurement diameter of 0.7310 inches.
  • Precise 0.7310 in measurement diameter
  • Constructed from steel
  • Class ZZ Minus tolerance
  • Tight tolerance of 0.000200 in