Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.7480 in Diameter, Black Oxide
Catalog No. 5000530077
Encompass
This Class ZZ Minus Plug & Pin Gage provides precise measurement with a 0.7480 in diameter. It features a durable black oxide finish and a tight tolerance of 0.000200 in, making it suitable for accurate gaging applications.
  • Precise diameter of 0.7480 in
  • Tight tolerance of 0.000200 in
  • Durable black oxide finish
  • Class ZZ minus type for specific gaging needs
Catalog No. 50-005-30077 Supplier MSC Supplier No. 56250970
This Class ZZ Minus Plug & Pin Gage provides precise measurement with a 0.7480 in diameter. It features a durable black oxide finish and a tight tolerance of 0.000200 in, making it suitable for accurate gaging applications.
  • Precise diameter of 0.7480 in
  • Tight tolerance of 0.000200 in
  • Durable black oxide finish
  • Class ZZ minus type for specific gaging needs