Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.7900 in Diameter, Black Oxide
Catalog No. 5000527934
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a diameter of 0.7900 inches and a tight tolerance of 0.000200 decimal inches, ensuring accuracy. The gage is finished with a durable black oxide coating.
  • Class ZZ Minus gage for accurate measurements
  • 0.7900 in diameter for specific sizing needs
  • 0.000200 in tolerance for high precision
  • Durable black oxide coating for enhanced protection
Catalog No. 50-005-27934 Supplier MSC Supplier No. 56251408
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a diameter of 0.7900 inches and a tight tolerance of 0.000200 decimal inches, ensuring accuracy. The gage is finished with a durable black oxide coating.
  • Class ZZ Minus gage for accurate measurements
  • 0.7900 in diameter for specific sizing needs
  • 0.000200 in tolerance for high precision
  • Durable black oxide coating for enhanced protection