Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.8180 in Diameter, Black Oxide Finish
Catalog No. 5000531775
Encompass
This plug and pin gage is designed for precise measurement applications, featuring a 0.8180 inch diameter and a durable black oxide finish. It is a Class ZZ Minus gage, suitable for verifying dimensions with high accuracy.
  • Diameter of measurement: 0.8180 inch
  • Tolerance: 0.000200 inch
  • Black oxide coating for enhanced durability
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-31775 Supplier MSC Supplier No. 56251689
This plug and pin gage is designed for precise measurement applications, featuring a 0.8180 inch diameter and a durable black oxide finish. It is a Class ZZ Minus gage, suitable for verifying dimensions with high accuracy.
  • Diameter of measurement: 0.8180 inch
  • Tolerance: 0.000200 inch
  • Black oxide coating for enhanced durability
  • Includes a certificate of accuracy with NIST traceability