Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.8880 in Diameter, Steel
Catalog No. 5000528709
Encompass
This plug and pin gage is designed for precise measurement applications, featuring a 0.8880 inch diameter. Manufactured from durable steel, it ensures accuracy with a Class ZZ minus tolerance of 0.000200 inch, making it suitable for demanding quality control and inspection tasks.
  • Designed for precise measurement applications.
  • Features a 0.8880 inch diameter for specific sizing requirements.
  • Constructed from durable steel for longevity and performance.
  • Maintains accuracy with a Class ZZ minus tolerance of 0.000200 "
  • Ideal for quality control and inspection tasks.
Catalog No. 50-005-28709 Supplier MSC Supplier No. 78380847
This plug and pin gage is designed for precise measurement applications, featuring a 0.8880 inch diameter. Manufactured from durable steel, it ensures accuracy with a Class ZZ minus tolerance of 0.000200 inch, making it suitable for demanding quality control and inspection tasks.
  • Designed for precise measurement applications.
  • Features a 0.8880 inch diameter for specific sizing requirements.
  • Constructed from durable steel for longevity and performance.
  • Maintains accuracy with a Class ZZ minus tolerance of 0.000200 "
  • Ideal for quality control and inspection tasks.