Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.8970 in Diameter, Steel
Catalog No. 5000528812
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Constructed from durable steel, it features a 0.8970 in diameter and is suitable for various industrial and laboratory settings requiring high accuracy.
  • Class ZZ Minus for precise gaging.
  • Constructed from steel for durability.
  • 0.8970 in diameter for specific measurement needs.
  • High tolerance of 0.000200 in.
Catalog No. 50-005-28812 Supplier MSC Supplier No. 78380938
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Constructed from durable steel, it features a 0.8970 in diameter and is suitable for various industrial and laboratory settings requiring high accuracy.
  • Class ZZ Minus for precise gaging.
  • Constructed from steel for durability.
  • 0.8970 in diameter for specific measurement needs.
  • High tolerance of 0.000200 in.