Learn More
MSC SPI Plug and Pin Gage, Class ZZ Minus, 0.9040 in, Steel
Catalog No. 5000531752
Encompass
This precision measuring tool is a Class ZZ Minus Plug and Pin Gage, featuring a specific diameter of 0.9040 inches. It offers a precise tolerance of 0.000200 decimal inches (0.0002 mm), ensuring high accuracy for various measurement applications. Constructed from durable steel, it is built for longevity and reliable performance in environments utilizing the decimal inch system of measurement.
  • Class ZZ Minus gage for precision measurement
  • Diameter of 0.9040 inches
  • Tolerance of 0.000200 decimal inches (0.0002 mm)
  • Made from durable steel
  • Suitable for applications using the decimal inch system
  • Provided in brand new condition
  • Includes a factory warranty
Catalog No. 50-005-31752 Supplier MSC Supplier No. 56252554
This precision measuring tool is a Class ZZ Minus Plug and Pin Gage, featuring a specific diameter of 0.9040 inches. It offers a precise tolerance of 0.000200 decimal inches (0.0002 mm), ensuring high accuracy for various measurement applications. Constructed from durable steel, it is built for longevity and reliable performance in environments utilizing the decimal inch system of measurement.
  • Class ZZ Minus gage for precision measurement
  • Diameter of 0.9040 inches
  • Tolerance of 0.000200 decimal inches (0.0002 mm)
  • Made from durable steel
  • Suitable for applications using the decimal inch system
  • Provided in brand new condition
  • Includes a factory warranty