Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.907 in Diameter, Black Oxide
Catalog No. 5000527932
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate measurement applications. It features a 0.907 inch diameter and a black oxide coating, offering a tolerance of 0.000200 inch (0.0002 mm).
  • Black oxide coating/finish
  • 0.000200 inch tolerance
  • 0.9070 inch diameter of measurement
Catalog No. 50-005-27932 Supplier MSC Supplier No. 56252588
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate measurement applications. It features a 0.907 inch diameter and a black oxide coating, offering a tolerance of 0.000200 inch (0.0002 mm).
  • Black oxide coating/finish
  • 0.000200 inch tolerance
  • 0.9070 inch diameter of measurement