Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.9390" Dia, Steel
Catalog No. 5000510145
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection applications. It offers high accuracy and durability for reliable gauging.
  • Class ZZ Minus for accurate gauging.
  • Diameter of 0.9390 inches for precise measurements.
  • Manufactured from steel for durability.
  • Tolerance of 0.000200 inches ensures high precision.
Catalog No. 50-005-10145 Supplier MSC Supplier No. 78381357
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection applications. It offers high accuracy and durability for reliable gauging.
  • Class ZZ Minus for accurate gauging.
  • Diameter of 0.9390 inches for precise measurements.
  • Manufactured from steel for durability.
  • Tolerance of 0.000200 inches ensures high precision.