Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.9440" Diameter, 0.0002" Tolerance, Black Oxide
Catalog No. 5000511640
Encompass
The SPI Class ZZ Minus Plug & Pin Gage is an essential tool for precision measurement applications. It features a specific diameter for gaging holes and ensuring tight tolerances, making it suitable for quality control and inspection tasks.
  • Designed for accurate measurement of bore diameters.
  • Ensures precise fit and tolerance checks.
  • Features a durable black oxide finish.
  • Meets Class ZZ Minus specifications.
  • Has a 0.9440 inch diameter.
  • Provides a precise 0.0002 inch tolerance.
Catalog No. 50-005-11640 Supplier MSC Supplier No. 78381407
The SPI Class ZZ Minus Plug & Pin Gage is an essential tool for precision measurement applications. It features a specific diameter for gaging holes and ensuring tight tolerances, making it suitable for quality control and inspection tasks.
  • Designed for accurate measurement of bore diameters.
  • Ensures precise fit and tolerance checks.
  • Features a durable black oxide finish.
  • Meets Class ZZ Minus specifications.
  • Has a 0.9440 inch diameter.
  • Provides a precise 0.0002 inch tolerance.