Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.9570" Dia
Catalog No. 5000516018
Encompass
This Class ZZ Minus Plug & Pin Gage measures 0.9570 inches in diameter and is constructed from steel. Designed for precision, it offers a tolerance of 0.000200 "
  • Measures 0.9570 inch in diameter
  • Tolerance of 0.000200 inch
  • Made from steel
  • Class ZZ Minus
Catalog No. 50-005-16018 Supplier MSC Supplier No. 89095707
This Class ZZ Minus Plug & Pin Gage measures 0.9570 inches in diameter and is constructed from steel. Designed for precision, it offers a tolerance of 0.000200 "
  • Measures 0.9570 inch in diameter
  • Tolerance of 0.000200 inch
  • Made from steel
  • Class ZZ Minus