Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.9990 inch Diameter, Steel
Catalog No. 5000531046
Encompass
This precise Class ZZ Minus Plug & Pin Gage is designed for accurate measurement, featuring a 0.9990 inch diameter.
  • Made from durable steel
  • Offers a precise tolerance of 0.000200 inch for accurate gauging
  • Manufactured to Class ZZ Minus standards
  • Ideal for measuring small holes and critical dimensions
Catalog No. 50-005-31046 Supplier MSC Supplier No. 78381951
This precise Class ZZ Minus Plug & Pin Gage is designed for accurate measurement, featuring a 0.9990 inch diameter.
  • Made from durable steel
  • Offers a precise tolerance of 0.000200 inch for accurate gauging
  • Manufactured to Class ZZ Minus standards
  • Ideal for measuring small holes and critical dimensions