Learn More
MSC SPI Plug Thread Gage, #0-80 Thread, 2B Class, Single End, Go
Catalog No. 5000537521
Encompass
This plug thread gage is designed for inspecting internal threads, specifically for #0-80 UNF threads with a 2B class. It features a single-end 'Go' design and is made from durable tool steel, ensuring precision and reliability in thread verification processes. Each gage includes an inspection certificate traceable to NIST standards (ASME B1.2) for quality assurance.
  • Used for inspection of internal threads to ensure assembly of mating components.
  • Made from durable tool steel.
  • Features a single-end 'Go' design for specific thread verification.
  • Includes an inspection certificate traceable to NIST standards (ASME B1.2).
  • Designed for #0-80 UNF thread size.
  • 2B class for accurate thread classification.
  • Taperlock design for secure handling.
Catalog No. 50-005-37521 Supplier MSC Supplier No. 75887430
This plug thread gage is designed for inspecting internal threads, specifically for #0-80 UNF threads with a 2B class. It features a single-end 'Go' design and is made from durable tool steel, ensuring precision and reliability in thread verification processes. Each gage includes an inspection certificate traceable to NIST standards (ASME B1.2) for quality assurance.
  • Used for inspection of internal threads to ensure assembly of mating components.
  • Made from durable tool steel.
  • Features a single-end 'Go' design for specific thread verification.
  • Includes an inspection certificate traceable to NIST standards (ASME B1.2).
  • Designed for #0-80 UNF thread size.
  • 2B class for accurate thread classification.
  • Taperlock design for secure handling.