Learn More
MSC SPI Plug Thread Gage, #0-80 Thread, 3B Class, Single End, No Go
Catalog No. 5000512870
Encompass
This plug thread gage is used for the inspection of internal threads to ensure the proper assembly of mating components. It features a #0-80 UNF thread, 3B class, and a right-hand thread direction, and is made from durable tool steel.
  • Used for inspection of internal threads to ensure assembly of mating components.
  • Made from steel.
  • Handle not included.
  • Traceability certification included: inspection certificate traceable to NIST standards: ASME B1.2.
  • UNF thread standard.
  • Right hand thread direction.
  • Single end.
  • No go type.
  • Overall length 0.8789 in.
Catalog No. 50-005-12870 Supplier MSC Supplier No. 75887455
This plug thread gage is used for the inspection of internal threads to ensure the proper assembly of mating components. It features a #0-80 UNF thread, 3B class, and a right-hand thread direction, and is made from durable tool steel.
  • Used for inspection of internal threads to ensure assembly of mating components.
  • Made from steel.
  • Handle not included.
  • Traceability certification included: inspection certificate traceable to NIST standards: ASME B1.2.
  • UNF thread standard.
  • Right hand thread direction.
  • Single end.
  • No go type.
  • Overall length 0.8789 in.