Learn More
MSC SPI Plug Thread Gage, #10-24 Thread, 2B Class, Single End, Go
Catalog No. 5000514206
Encompass
This plug thread gage is designed for the inspection of internal threads to ensure the proper assembly of mating components.
  • Taperlock
  • Inspection of internal threads to ensure assembly of mating components
Catalog No. 50-005-14206 Supplier MSC Supplier No. 75887877
This plug thread gage is designed for the inspection of internal threads to ensure the proper assembly of mating components.
  • Taperlock
  • Inspection of internal threads to ensure assembly of mating components