Learn More
MSC SPI Plug Thread Gage, #12-28, Class 3B, Double End Go/No Go
Catalog No. 5000530273
Encompass
This plug thread gage is designed for measuring #12-28 threads. It features a double-end design with both "Go" and "No Go" functions, and includes a handle for ease of use.
  • Designed for measuring #12-28 threads.
  • Features a 3B class for precise measurements.
  • Double-end design includes both "Go" and "No Go" functions.
  • Constructed from durable steel material.
  • Handle is included for convenient operation.
  • NIST traceability certification included.
  • Manufactured in India.
Catalog No. 50-005-30273 Supplier MSC Supplier No. 75889246
This plug thread gage is designed for measuring #12-28 threads. It features a double-end design with both "Go" and "No Go" functions, and includes a handle for ease of use.
  • Designed for measuring #12-28 threads.
  • Features a 3B class for precise measurements.
  • Double-end design includes both "Go" and "No Go" functions.
  • Constructed from durable steel material.
  • Handle is included for convenient operation.
  • NIST traceability certification included.
  • Manufactured in India.