Learn More
MSC SPI Plug Thread Gage, 3/8-32 Thread, 2B Class, No Go, Single End
Catalog No. 5000533019
Encompass
This plug thread gage is designed for the inspection of internal threads to ensure the proper assembly of mating components. It features a 3/8-32 UNEF thread with a 2B class, No Go designation, and is made from durable tool steel.
  • Taperlock design
  • Designed for inspection of internal threads
  • Ensures assembly of mating components
  • Single end configuration
  • Right hand thread direction
  • Manufactured to ASME B1.2 standards
  • Traceability certification included, traceable to NIST standards
  • Made from tool steel
Catalog No. 50-005-33019 Supplier MSC Supplier No. 55307797
This plug thread gage is designed for the inspection of internal threads to ensure the proper assembly of mating components. It features a 3/8-32 UNEF thread with a 2B class, No Go designation, and is made from durable tool steel.
  • Taperlock design
  • Designed for inspection of internal threads
  • Ensures assembly of mating components
  • Single end configuration
  • Right hand thread direction
  • Manufactured to ASME B1.2 standards
  • Traceability certification included, traceable to NIST standards
  • Made from tool steel