Learn More
MSC SPI Plug Thread Gage, 7/16-20 Thread, 2B Class, Double End, Go & No Go
Catalog No. 5000513083
Encompass
This thread plug gage is designed for accurately checking internal threads. It features a double-ended design with both 'Go' and 'No Go' sides, allowing for quick verification of thread conformity. Constructed from durable steel, it ensures reliable performance and includes a handle for ease of use. A NIST traceability certification is provided, confirming its adherence to measurement standards.
  • Go and No Go functionality
  • Checks 7/16-20 thread size
  • Constructed from steel
  • Features 2B classification
  • Includes a handle for convenient use
  • NIST traceability certification included
  • Double-ended design
Catalog No. 50-005-13083 Supplier MSC Supplier No. 75889451
This thread plug gage is designed for accurately checking internal threads. It features a double-ended design with both 'Go' and 'No Go' sides, allowing for quick verification of thread conformity. Constructed from durable steel, it ensures reliable performance and includes a handle for ease of use. A NIST traceability certification is provided, confirming its adherence to measurement standards.
  • Go and No Go functionality
  • Checks 7/16-20 thread size
  • Constructed from steel
  • Features 2B classification
  • Includes a handle for convenient use
  • NIST traceability certification included
  • Double-ended design