Learn More
MSC SPI Plug Thread Gage, 9/16-18 Thread, 2B Class, Single End, No Go
Catalog No. 5000531426
Encompass
This plug thread gage is designed for inspecting internal threads to ensure the proper assembly of mating components. It features a 9/16-18 thread size and 2B class, with a single-end, no-go configuration.
  • Taperlock
  • Inspection of internal threads to ensure assembly of mating components
Catalog No. 50-005-31426 Supplier MSC Supplier No. 75888487
This plug thread gage is designed for inspecting internal threads to ensure the proper assembly of mating components. It features a 9/16-18 thread size and 2B class, with a single-end, no-go configuration.
  • Taperlock
  • Inspection of internal threads to ensure assembly of mating components