Learn More
MSC SPI Plug Thread Gage Handle, 4 Handle Size, Steel
Catalog No. 5000530705
Encompass
This plug thread gage handle is designed to securely hold plug gages for various thread sizes. It features a Taper Lock system that ensures a snug fit by using the interference between mating surfaces, keeping the gage tightly in place during use.
  • Handle size 4
  • Constructed from steel for durability
  • Secure taper lock mechanism for holding gages
  • Hex dimension of 0.862 in
  • Overall length of 3.625 in
  • Side hole diameter of 0.375 in
  • Through hole ID at ends is 0.605 in
  • No tapped hole
  • Certified to NIST and ANSI standards
Catalog No. 50-005-30705 Supplier MSC Supplier No. 75890103
This plug thread gage handle is designed to securely hold plug gages for various thread sizes. It features a Taper Lock system that ensures a snug fit by using the interference between mating surfaces, keeping the gage tightly in place during use.
  • Handle size 4
  • Constructed from steel for durability
  • Secure taper lock mechanism for holding gages
  • Hex dimension of 0.862 in
  • Overall length of 3.625 in
  • Side hole diameter of 0.375 in
  • Through hole ID at ends is 0.605 in
  • No tapped hole
  • Certified to NIST and ANSI standards