Learn More
MSC SPI Plug Thread Gage, M20x1.5 Thread, 6H Class, No Go, Single End
Catalog No. 5000535136
Encompass
This plug thread gage is designed for the precise inspection of internal threads, ensuring proper assembly of mating components. It features an M20x1.5 thread size with a 6H class and is a single-ended, no-go type gage.
  • Taperlock design
  • Inspects internal threads to ensure assembly of mating components
  • No Go functionality for precise measurement
  • M20x1.5 thread size
  • 6H class for metric threads
  • Constructed from tool steel
  • Right hand thread direction
  • Metric thread standard
  • Single end design
Catalog No. 50-005-35136 Supplier MSC Supplier No. 54335302
This plug thread gage is designed for the precise inspection of internal threads, ensuring proper assembly of mating components. It features an M20x1.5 thread size with a 6H class and is a single-ended, no-go type gage.
  • Taperlock design
  • Inspects internal threads to ensure assembly of mating components
  • No Go functionality for precise measurement
  • M20x1.5 thread size
  • 6H class for metric threads
  • Constructed from tool steel
  • Right hand thread direction
  • Metric thread standard
  • Single end design