Learn More
MSC SPI Plug Thread Gage, M3x0.5, 6H Class, Go, Tool Steel
Catalog No. 5000532457
Encompass
This Plug Thread Gage is designed for inspecting internal M3x0.5 threads with a 6H class. Made from tool steel and through hardened to RC 60-64, this single-end, Go gage ensures assembly of mating components by verifying thread measurements according to ANSI specification 1.16B. It includes an inspection certificate traceable to NIST standards (ASME B1.16M).
  • Taperlock design
  • Inspects internal threads to ensure assembly of mating components
  • Through hardened to RC 60-64
  • Thread measurements as per ANSI specification 1.16B
  • Includes certificate of calibration traceable to NIST
  • Designed for M3x0.5 threads
  • 6H class
  • Right hand thread direction
  • Single end configuration
Catalog No. 50-005-32457 Supplier MSC Supplier No. 54334321
This Plug Thread Gage is designed for inspecting internal M3x0.5 threads with a 6H class. Made from tool steel and through hardened to RC 60-64, this single-end, Go gage ensures assembly of mating components by verifying thread measurements according to ANSI specification 1.16B. It includes an inspection certificate traceable to NIST standards (ASME B1.16M).
  • Taperlock design
  • Inspects internal threads to ensure assembly of mating components
  • Through hardened to RC 60-64
  • Thread measurements as per ANSI specification 1.16B
  • Includes certificate of calibration traceable to NIST
  • Designed for M3x0.5 threads
  • 6H class
  • Right hand thread direction
  • Single end configuration