Learn More
MSC SPI Plug Thread Gage, M5x0.8 Thread, 6H Class, Single End, Go, Tool Steel
Catalog No. 5000527098
Encompass
This plug thread gage is designed for inspecting internal M5x0.8 threads. It features a 6H class and is a single-end, go gage. Made from durable tool steel, it ensures the proper fit and assembly of mating components.
  • Taperlock design
  • Allows for inspection of internal threads to ensure assembly of mating components
  • Handle not included
Catalog No. 50-005-27098 Supplier MSC Supplier No. 54334271
This plug thread gage is designed for inspecting internal M5x0.8 threads. It features a 6H class and is a single-end, go gage. Made from durable tool steel, it ensures the proper fit and assembly of mating components.
  • Taperlock design
  • Allows for inspection of internal threads to ensure assembly of mating components
  • Handle not included