Learn More
MSC SPI Plug Thread Gage, M6x1 Thread, 6H Class, Single End, Go
Catalog No. 5000535162
Encompass
This plug thread gage is designed for the inspection of internal threads to ensure the proper assembly of mating components. It features an M6x1 thread size with a 6H classification and is a single-end, 'Go' type gage. Constructed from durable tool steel, it is right-hand threaded and comes with an inspection certificate for traceability.
  • Taperlock design
  • Inspects internal threads to ensure assembly of mating components
  • M6x1 thread size
  • 6H class for precision
  • Single end 'Go' type
  • Made from tool steel
  • Right-hand thread direction
  • Includes inspection certificate
Catalog No. 50-005-35162 Supplier MSC Supplier No. 54334289
This plug thread gage is designed for the inspection of internal threads to ensure the proper assembly of mating components. It features an M6x1 thread size with a 6H classification and is a single-end, 'Go' type gage. Constructed from durable tool steel, it is right-hand threaded and comes with an inspection certificate for traceability.
  • Taperlock design
  • Inspects internal threads to ensure assembly of mating components
  • M6x1 thread size
  • 6H class for precision
  • Single end 'Go' type
  • Made from tool steel
  • Right-hand thread direction
  • Includes inspection certificate