Learn More
MSC SPI Plug Thread Gage, M7x1 Thread, 6H Class, Single End, No Go
Catalog No. 5000527090
Encompass
This plug thread gage is an essential tool for verifying internal threads, ensuring accurate assembly of mating components. It is designed with a taperlock mechanism and functions as a No Go gage, specifically for M7x1 threads of 6H class in a single-end configuration.
  • Taperlock design
  • Ensures proper inspection of internal threads
  • No Go gage type
  • M7x1 thread size
  • 6H class
  • Single-end configuration
  • Constructed from tool steel
  • Includes inspection certificate traceable to NIST
  • Complies with ASME B1.16M standards
Catalog No. 50-005-27090 Supplier MSC Supplier No. 54335070
This plug thread gage is an essential tool for verifying internal threads, ensuring accurate assembly of mating components. It is designed with a taperlock mechanism and functions as a No Go gage, specifically for M7x1 threads of 6H class in a single-end configuration.
  • Taperlock design
  • Ensures proper inspection of internal threads
  • No Go gage type
  • M7x1 thread size
  • 6H class
  • Single-end configuration
  • Constructed from tool steel
  • Includes inspection certificate traceable to NIST
  • Complies with ASME B1.16M standards