Learn More
MSC SPI Plug Thread Gage, No Go, #6-32 Thread, 2B Class, Single End
Catalog No. 5000512655
Encompass
This plug thread gage is designed for inspecting internal threads. It features a #6-32 thread, 2B class, and is a single-end, no-go type. Constructed from tool steel, it requires a size 00 handle (not included).
  • Taperlock
  • Inspection of internal threads to ensure assembly of mating components
Catalog No. 50-005-12655 Supplier MSC Supplier No. 75887760
This plug thread gage is designed for inspecting internal threads. It features a #6-32 thread, 2B class, and is a single-end, no-go type. Constructed from tool steel, it requires a size 00 handle (not included).
  • Taperlock
  • Inspection of internal threads to ensure assembly of mating components