Learn More
MSC SPI Point Micrometers, Electronic, Ratchet Thimble, 1-2 in, ±.0002 in Accuracy
Catalog No. 5000536608
Encompass
These electronic point micrometers are designed for precise measurements, offering a range of 1 to 2 inches. They feature a ratchet thimble and a digital counter for accurate readings. With a throat depth of 44.5 mm, these micrometers provide an accuracy of ±.0002 inches.
  • Electronic operation
  • Ratchet thimble
  • Digital counter for clear readings
  • Positive lock nut for secure measurements
  • Carbide anvil for durability
  • Rotating spindle
  • Data output capability
  • Includes a plastic box for storage
  • Calibrated for accuracy
  • SUS420F spindle material
  • High resolution of 0.000
  • 6.5 mm spindle diameter
  • Fine graduation of 0.0001
  • 5 mm spindle and anvil point length
  • 30-degree spindle and anvil point angle
  • Comes with one 3V CR2032 lithium battery
  • Complies with ASME 89.1.13 standards
Catalog No. 50-005-36608 Supplier MSC Supplier No. 13435276
These electronic point micrometers are designed for precise measurements, offering a range of 1 to 2 inches. They feature a ratchet thimble and a digital counter for accurate readings. With a throat depth of 44.5 mm, these micrometers provide an accuracy of ±.0002 inches.
  • Electronic operation
  • Ratchet thimble
  • Digital counter for clear readings
  • Positive lock nut for secure measurements
  • Carbide anvil for durability
  • Rotating spindle
  • Data output capability
  • Includes a plastic box for storage
  • Calibrated for accuracy
  • SUS420F spindle material
  • High resolution of 0.000
  • 6.5 mm spindle diameter
  • Fine graduation of 0.0001
  • 5 mm spindle and anvil point length
  • 30-degree spindle and anvil point angle
  • Comes with one 3V CR2032 lithium battery
  • Complies with ASME 89.1.13 standards