Learn More
MSC SPI Radius Gage, 3/8 Inch, Satin Chrome Finish, Steel
Catalog No. 5000529689
Encompass
The SPI 3/8 Inch Radius Gage is a precision measuring tool designed for checking both convex and concave radii. It features a satin chrome finish and is constructed from steel, offering durability and accuracy for various applications.
  • Measures 3/8 inch radius.
  • Convex and concave radii per each leaf.
  • Accuracy of ±0.0012 "
  • Satin chrome finish for enhanced visibility and protection.
  • Constructed from durable steel.
  • Ideal for quality control and inspection tasks.
Catalog No. 50-005-29689 Supplier MSC Supplier No. 01718964
The SPI 3/8 Inch Radius Gage is a precision measuring tool designed for checking both convex and concave radii. It features a satin chrome finish and is constructed from steel, offering durability and accuracy for various applications.
  • Measures 3/8 inch radius.
  • Convex and concave radii per each leaf.
  • Accuracy of ±0.0012 "
  • Satin chrome finish for enhanced visibility and protection.
  • Constructed from durable steel.
  • Ideal for quality control and inspection tasks.